missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH1A2 Recombinant Rabbit Monoclonal Antibody (23GB3640), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552583
This item is not returnable.
View return policy
Description
This protein belongs to the aldehyde dehydrogenase family of proteins. The product of this gene is an enzyme that catalyzes the synthesis of retinoic acid (RA) from retinaldehyde. Retinoic acid, the active derivative of vitamin A (retinol), is a hormonal signaling molecule that functions in developing and adult tissues. The studies of a similar mouse gene suggest that this enzyme and the cytochrome CYP26A1, concurrently establish local embryonic retinoic acid levels which facilitate posterior organ development and prevent spina bifida. Three transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| ALDH1A2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ALDH1A2 | |
| AL1A2; alcohol dehydrogenase family 1, subfamily A2; alcohol dehydrogenase family 1, subfamily A7; aldehyde dehydrogenase 1 family member A2; aldehyde dehydrogenase 1 family, member A2; aldehyde dehydrogenase 1A2; aldehyde dehydrogenase family 1 member A2; aldehyde dehydrogenase family 1, subfamily A2; Aldh1a2; Aldh1a7; AV116159; fb50h01; neckless; nls; nof; no-fin; RALDH 2; RALDH(II); Raldh1; RALDH2; Raldh-2; RALDH2-T; retinal dehydrogenase 2; retinal dehydrogenase, type II; retinaldehyde dehydrogenase 2; retinaldehyde dehydrogenase type 2; retinaldehyde-specific dehydrogenase type 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8854 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3640 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O94788 | |
| ALDH1A2 | |
| A synthetic peptide of human ALDH1A2 (5-50AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction