missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH16A1 Recombinant Rabbit Monoclonal Antibody (23GB4315), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552682
This item is not returnable.
View return policy
Description
This gene encodes a member of the aldehyde dehydrogenase superfamily. The family members act on aldehyde substrates and use nicotinamide adenine dinucleotide phosphate (NADP) as a cofactor. This gene is conserved in chimpanzee, dog, cow, mouse, rat, and zebrafish. The protein encoded by this gene interacts with maspardin, a protein that when truncated is responsible for Mast syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| ALDH16A1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ALDH16A1 | |
| 2410004H02Rik; AI452234; aldehyde dehydrogenase 16 family member A1; aldehyde dehydrogenase 16 family, member A1; aldehyde dehydrogenase 16A1; aldehyde dehydrogenase family 16 member A1; ALDH16A1; LOW QUALITY PROTEIN: aldehyde dehydrogenase family 16 member A1; mFLJ00209 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 126133, 361571, 69748 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4315 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3T1L0, Q571I9, Q8IZ83 | |
| ALDH16A1 | |
| A synthesized peptide derived from human ALDH16A1 (550-590AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction