missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH16A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545982
This item is not returnable.
View return policy
Description
Peptide sequence: YGPVPESHAC ALAWLDTQDR CLGHYVNGKW LKPEHRNSVP CQDPITGENL Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 92%; Rabbit: 79%; Rat: 100%.
This gene encodes a member of the aldehyde dehydrogenase superfamily. The family members act on aldehyde substrates and use nicotinamide adenine dinucleotide phosphate (NADP) as a cofactor. This gene is conserved in chimpanzee, dog, cow, mouse, rat, and zebrafish. The protein encoded by this gene interacts with maspardin, a protein that when truncated is responsible for Mast syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| ALDH16A1 | |
| Polyclonal | |
| Unconjugated | |
| ALDH16A1 | |
| 2410004H02Rik; AI452234; aldehyde dehydrogenase 16 family member A1; aldehyde dehydrogenase 16 family, member A1; aldehyde dehydrogenase 16A1; aldehyde dehydrogenase family 16 member A1; ALDH16A1; LOW QUALITY PROTEIN: aldehyde dehydrogenase family 16 member A1; mFLJ00209 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 126133 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IZ83 | |
| ALDH16A1 | |
| Synthetic peptide directed towards the N-terminal of human ALDH16A1 (aa 19-68). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction