missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldehyde Oxidase 1/AOX1 Antibody (PSH0-17), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32018
This item is not returnable.
View return policy
Description
Aldehyde Oxidase 1/AOX1 Monoclonal antibody specifically detects Aldehyde Oxidase 1/AOX1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Aldehyde Oxidase 1/AOX1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| aldehyde oxidase, aldehyde oxidase 1, AOAOH1, EC 1.2.3.1 | |
| Synthetic peptide within human Aldehyde Oxidase 1/AOX1 aa 1,301-1,338/ 1,338. (Uniprot: Q06278) | |
| 100 μL | |
| Cancer, Endocrinology, Signal Transduction | |
| 316 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| PSH0-17 | |
| Unconjugated | |
| PBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction