missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldehyde Dehydrogenase 3-A1/ALDH3A1 Antibody (OTI1B6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70152
This item is not returnable.
View return policy
Description
Aldehyde Dehydrogenase 3-A1/ALDH3A1 Monoclonal specifically detects Aldehyde Dehydrogenase 3-A1/ALDH3A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications
| Aldehyde Dehydrogenase 3-A1/ALDH3A1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, CyTOF-ready | |
| Aldehyde dehydrogenase 3, aldehyde dehydrogenase 3 family, member A1, Aldehyde dehydrogenase family 3 member A1, aldehyde dehydrogenase isozyme 3, aldehyde dehydrogenase type III, ALDH3aldehyde dehydrogenase, dimeric NADP-preferring, ALDHIII, EC 1.2.1, EC 1.2.1.5, MGC10406, stomach aldehyde dehydrogenase | |
| Mouse | |
| 50.2 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF | |
| OTI1B6 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| ALDH3A1 | |
| Full length human recombinant protein of human ALDH3A1(NP_000682) produced in HEK293T cell. | |
| Affinity Purified | |
| Vision | |
| 218 | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction