missing translation for 'onlineSavingsMsg'
Learn More

Aldehyde Dehydrogenase 1-A1/ALDH1A1 Antibody (ALDH1A1/7011R), DyLight 550, Novus Biologicals™

Product Code. 18377975 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377975 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377975 Supplier Novus Biologicals Supplier No. NBP314211R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Aldehyde Dehydrogenase 1-A1/ALDH1A1 Monoclonal antibody specifically detects Aldehyde Dehydrogenase 1-A1/ALDH1A1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aldehyde Dehydrogenase 1-A1/ALDH1A1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ALDH1A1/7011R
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias acetaldehyde dehydrogenase 1, ALDC, aldehyde dehydrogenase 1 family, member A1, aldehyde dehydrogenase 1, soluble, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic, aldehyde dehydrogenase, liver cytosolic, ALDH class 1, ALDH1, ALDH11, ALDH-E1, ALHDII, EC 1.2.1, EC 1.2.1.36, MGC2318, PUMB1, RALDH 1, RalDH1, retinal dehydrogenase 1, retinaldehyde dehydrogenase 1
Host Species Rabbit
Immunogen Recombinant fragment (around aa 315-434) of human Aldehyde Dehydrogenase 1-A1/ALDH1A1 protein (exact sequence is proprietary). (Uniprot: P00352 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 216
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.