missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldehyde Dehydrogenase 1-A1/ALDH1A1 Antibody (8F1-R), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-32019
This item is not returnable.
View return policy
Description
Aldehyde Dehydrogenase 1-A1/ALDH1A1 Monoclonal antibody specifically detects Aldehyde Dehydrogenase 1-A1/ALDH1A1 in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Aldehyde Dehydrogenase 1-A1/ALDH1A1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| acetaldehyde dehydrogenase 1, ALDC, aldehyde dehydrogenase 1 family, member A1, aldehyde dehydrogenase 1, soluble, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic, aldehyde dehydrogenase, liver cytosolic, ALDH class 1, ALDH1, ALDH11, ALDH-E1, ALHDII, EC 1.2.1, EC 1.2.1.36, MGC2318, PUMB1, RALDH 1, RalDH1, retinal dehydrogenase 1, retinaldehyde dehydrogenase 1 | |
| Recombinant protein within Human Aldehyde Dehydrogenase 1-A1/ALDH1A1 aa 1-501/ 501. (Uniprot: P00352) | |
| 100 μL | |
| Cancer, Diabetes Research, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers | |
| 216 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 8F1-R | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction