missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ALDEFLUOR™ DEAB Reagent, 1.5 mM, 1 mL

Product Code. 17189991
Encompass_Preferred
Change view
Click to view available options
:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17189991 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17189991 Supplier STEMCELL Technologies Supplier No. 01705

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of ALDH enzyme used as a control in the ALDEFLUOR™ assay system.

  • The ALDEFLUOR™ reagent system is a non-immunological method to identify stem/progenitor cells on the basis of their aldehyde dehydrogenase (ALDH) activity
  • High ALDH expression has been reported for normal and cancer precursor cells of various lineages, including hematopoietic, mammary, endothelial, mesenchymal, and neural
  • Cells expressing high levels of ALDH become brightly fluorescent (ALDHbr) and can be identified and enumerated using a standard flow cytometer or isolated by cell sorting for further purification and characterization
  • The use of an internal cellular enzyme for identifying and isolating stem and progenitor cells provides an alternative to the more traditional method of staining with antibodies against cell surface antigens

Specifications

CAS 64-17-5
Cell Type Brain Tumor Stem Cells; Cancer Cells and Cell Lines; Hematopoietic Stem and Progenitor Cells; Mammary Cells; Mesenchymal Stem and Progenitor Cells; Neural Stem and Progenitor Cells; Other
For Use With (Application) Flow Cytometry
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.