missing translation for 'onlineSavingsMsg'
Learn More

VEE GEE Alcohol Hydrometer with Certification

Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.

Brand:  VEE GEE 6613-K-C

Additional Details : Weight : 0.18000kg

 View more versions of this product

Product Code. 17826483

  • £419.00 / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Description

Description

  • Alcohol proof range differ depending on model
  • IRS Specification Size differ depending on model
  • li>Calibration for use at 15.5°C/60°F
  • Approx. Length: 305 mm
Specifications

Specifications

Alcohol Hydrometer
30.5 cm
15.5°C (60°F)
0.20%
Alcohol Hydrometer, Proof Scale, IRS Specification, Size K, with Certification
75 to 95%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers