Learn More
VEE GEE Alcohol Hydrometer
Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.
£62.00 - £66.15
Product Code | Brand | Alcohol Range | Increments | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Alcohol Range | Increments | Price | Quantity & Availability | ||||
17866523
|
VEE GEE
6613-Q |
165% to 185% | 0.2%/±0.2% |
£66.15
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
17896553
|
VEE GEE
6613-I |
60 to 80% | 0.2%/±0.3% |
£66.15
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
17826563
|
VEE GEE
6613-F |
0% to 20% | 0.2%/±0.3% |
£66.15
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
17896563
|
VEE GEE
6613-K |
75% to 95% | 0.2%/±0.3% |
£66.15
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
17896523
|
VEE GEE
6613-B |
80 to 120% | 0.2%/±0.3% |
£62.00
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
17836563
|
VEE GEE
6613-H |
40 to 60% | 0.2%/±0.4% |
£66.15
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
17806573
|
VEE GEE
6613-N |
125% to 145% | 0.2%/±0.3% |
£66.15
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
17886563
|
VEE GEE
6613-L |
90 to 110% | 0.2%/±0.3% |
£66.15
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
17886613
|
VEE GEE
6613-M |
105% to 125% | 0.2%/±0.3% |
£66.15
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
17886473
|
VEE GEE
6613-R |
185 to 206% | 0.2%/±0.2% |
£66.15
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
17876473
|
VEE GEE
6613-P |
145% to 165% | 0.2%/±0.3% |
£66.15
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
17836473
|
VEE GEE
6613-G |
20 to 40% | 0.2%/±0.4% |
£66.15
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
Description
- Alcohol proof range differ depending on model
- IRS Specification Size differ depending on model li>Calibration for use at 15.5°C/60°F
- Approx. Length: 305 mm
Specifications
Alcohol Hydrometer | |
Glass |