missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ alcohol dehydrogenase Antibody (SR3872), Novus Biologicals™
Rabbit Monoclonal Antibody
£240.00 - £290.00
Specifications
| Antigen | alcohol dehydrogenase |
|---|---|
| Clone | SR3872 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041794
|
Novus Biologicals™
NBP3-27812-100ul |
100 μL |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041567
|
Novus Biologicals™
NBP3-27812-50ul |
50 μL |
£240.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alcohol dehydrogenase Monoclonal antibody specifically detects alcohol dehydrogenase in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| alcohol dehydrogenase | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ADH, alpha subunit, ADH1alcohol dehydrogenase 1A, alcohol dehydrogenase 1 (class I), alpha polypeptide, alcohol dehydrogenase 1A (class I), alpha polypeptide, Alcohol dehydrogenase subunit alpha, aldehyde reductase, EC 1.1.1, EC 1.1.1.1 | |
| A synthesized peptide derived from human alcohol dehydrogenase (Uniprot #: P07327) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3872 | |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Diabetes Research, Lipid and Metabolism | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 124 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title