missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alcohol dehydrogenase 7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-90232
This item is not returnable.
View return policy
Description
alcohol dehydrogenase 7 Polyclonal specifically detects alcohol dehydrogenase 7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| alcohol dehydrogenase 7 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| ADH4, ADH-4, alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide, alcohol dehydrogenase class 4 mu/sigma chain, Alcohol dehydrogenase class IV mu/sigma chain, alcohol dehydrogenase VII, alcohol dehydrogenase-7, class IV sigma-1 alcohol dehydrogenase, class IV sigmasigma alcohol dehydrogenase, EC 1.1.1, EC 1.1.1.1, Gastric alcohol dehydrogenase, Retinol dehydrogenase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 131 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ADH7 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KFEKAMAVGATECISPKDSTKPISEVLSEMTGNNVGYTFEVIGHLETMIDALASCHMNYGTSV | |
| 0.1 mL | |
| Primary | |
| Specificity of human alcohol dehydrogenase 7 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction