missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ alcohol dehydrogenase 4 293T Cell Transient Overexpression Lysate
Shop All Bio Techne ProductsSpecifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Lipid and Metabolism |
| Target | alcohol dehydrogenase 4 |
| Synonym | ADH-2, alcohol dehydrogenase 4, alcohol dehydrogenase 4 (class II), pi polypeptide, Alcohol dehydrogenase class II pi chain, aldehyde reductase, EC 1.1.1, EC 1.1.1.1 |
| Gene Symbol | ADH4 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?