missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alcohol dehydrogenase 1A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
alcohol dehydrogenase 1A Polyclonal specifically detects alcohol dehydrogenase 1A in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | alcohol dehydrogenase 1A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | ADH, alpha subunit, ADH1alcohol dehydrogenase 1A, alcohol dehydrogenase 1 (class I), alpha polypeptide, alcohol dehydrogenase 1A (class I), alpha polypeptide, Alcohol dehydrogenase subunit alpha, aldehyde reductase, EC 1.1.1, EC 1.1.1.1 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human alcohol dehydrogenase 1A. Peptide sequence PESNYCLKNDVSNPQGTLQDGTSRFTCRRKPIHHFLGISTFSQYTVVDEN |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?