missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alcohol dehydrogenase 1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | alcohol dehydrogenase 1A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
alcohol dehydrogenase 1A Polyclonal specifically detects alcohol dehydrogenase 1A in Human samples. It is validated for Western Blot.Specifications
| alcohol dehydrogenase 1A | |
| Polyclonal | |
| Rabbit | |
| P07327 | |
| 124 | |
| Synthetic peptides corresponding to ADH1A(alcohol dehydrogenase 1A (class I), alpha polypeptide) The peptide sequence was selected from the N terminal of ADH1A (NP_000658). Peptide sequence: ESNYCLKNDVSNPQGTLQDGTSRFTCRRKPIHHFLGISTFSQYTVVDENA. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| ADH, alpha subunit, ADH1alcohol dehydrogenase 1A, alcohol dehydrogenase 1 (class I), alpha polypeptide, alcohol dehydrogenase 1A (class I), alpha polypeptide, Alcohol dehydrogenase subunit alpha, aldehyde reductase, EC 1.1.1, EC 1.1.1.1 | |
| ADH1A | |
| IgG | |
| 40 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title