Learn More
Alcohol Aldehyde-Free According to European Pharmacopoeia (EP), Reagecon
High-purity, stable product for European Pharmacopeia applications.
Brand: Reagecon 1002501
Code : A0
Additional Details : CAS Number : 64-17-5 Weight : 0.81000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Description
- European Pharmacopoeia Reagent, Solution
- Aldehyde-free
- High purity
- Accurate, stable and reproducible
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Developed and tested for European Pharmacopoeia (EP) applications.
- For preparation of other reagents (4.1.1), use in volumetric reagents (4.2.2) and used as a reagent in the performance of tests outlined in several EP monographs.
Specifications
1 L | |
EP Testing | |
Liquid |
Safety and Handling
- ALCOHOL ALDEHYDE FREE EUROPEAN PHARMACOPOEIA
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol