missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Alcian Blue/PAS Special Stain Kit

Product Code. 12523667 Shop All Epredia Products
Click to view available options
Type:
Alcian Blue
Alcian Blue/PAS Special Stain Kit
Periodic Acid Schiff
Unit Size:
500mL
Pack of 3
This item is not returnable. View return policy

Product Code. 12523667

Brand: Epredia™ 88043

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Demonstrate the presence of both neutral and acidic mucosubstances with the Epredia™ Alcian Blue/PAS Special Stain Kit

  • Can be used alone to stain acid mucins and other carboxylated and sulfated acid mucosubstances blue on tissue sections
  • Periodic Acid Schiff (PAS) reaction is used to stain glycogen, basement membranes and neutral mucosubstances

Includes: Alcian Blue pH 2.5 stain solution, periodic acid solution (0.5%) and Schiff reagent

TRUSTED_SUSTAINABILITY

Specifications

Type Alcian Blue
compliance-icons compliance-icons
Produktbeteckning
  • Alcian Blue pH 2.5 Stain Solution
Signalord
  • Danger
Riskkategorier
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Faroangivelser
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
Skyddsangivelse
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P364-And wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Kompletterande information
  • MIXTURE LIST-Contains : 2-methyl-3-isothiazolone, acetic acid, 5-chloro-2-methyl-3-isothiazolone, copper(IV) [[N,N',N'',N'''
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.