missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALCAM/CD166 Rabbit anti-Mouse, Clone: 311, Novus Biologicals™
Rabbit Monoclonal Antibody
£429.00
Specifications
| Antigen | ALCAM/CD166 |
|---|---|
| Clone | 311 |
| Dilution | Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
Description
ALCAM/CD166 Monoclonal specifically detects ALCAM/CD166 in Mouse samples. It is validated for Flow Cytometry, ELISA.Specifications
| ALCAM/CD166 | |
| Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
| Recombinant Mouse ALCAM/CD166 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 311 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 214 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title