missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alcaftadine, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Alcaftadine (R89674) is a histamine H1 receptor antagonist, which is used to prevent eye irritation brought on by allergic conjunctivitis. Alcaftadine is a broad-spectrum antihistamine displaying a high affinity for histamine H1 and H2 receptors and a lower affinity for H4 receptors. Alcaftadine also exhibits modulatory action on immune cell recruitment and mast cell stabilizing effects.
Specifications
Specifications
| Chemical Name or Material | Alcaftadine |
| CAS | 147084-10-4 |
| Color | Earth Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C19H21N3O |
| Quantity | 10 mg |
| Synonym | R89674 |
| Solubility Information | DMSO : ≥ 40 mg/mL (130.13 mM) |
| SMILES | O=CC1=CN=C2N1CCC3=CC=CC=C3/C2=C4CCN(C)CC\4 |
| Molecular Weight (g/mol) | 307.39 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction