missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Albuterol Sulfate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Salbutamol sulfate |
| CAS | 51022-70-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | 2 C13 H21 N O3 . H2 O4 S |
| Quantity | 1 g |
| Synonym | Salbutamol sulphate, Albuterol sulphate, 1,3-Benzenedimethanol, α1-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-, sulfate (2:1), 1,3-Benzenedimethanol, α1-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-, sulfate (2:1) (salt) (9CI), (±)-Salbutamol sulfate, Aerolin, Airet, Albuterol hemisulfate, Albuterol sulfate, Almotex, Anebron, Asmadil, Asmalin, Asmanil, Asmasal, Asmatol, Asmaven, Asmavent, Asmidon, Asmol Uni-Dose, Asthalin, Broncho-Spray, Broncovaleas, Bronter, Bugonol, Butamol, Buto-Asma, Butotal, Buventol, Cetsim, Cobutolin, Dilatamol, Ecovent, Farcolin, Grafalin, Instavent, Libretin, Loftan, Medolin, Mozal, NSC 289928, Novosalmol, Parasma, Proventil, Respax, Salbetol, Salbovent, Salbron, Salbulin, Salbumol, Salbusian, Salbutalan, Salbutamol hemisulfate, Salbutamol sulfate, Salbutan, Salbutard, Salbutine, Salbutol, Salbuven, Salbuvent, Salmaplon, Salomol, Salorain, Sch 13949W sulfate, Sedalin, Suprasma, Theosal, Tobybron, Vencronyl, Venetlin, Ventelin, Ventilan, Ventimax, Ventodiscks, Ventolin Rotacaps, Ventorain, Ventorline CR, Volma, dl-Salbutamol sulfate |
| InChI Formula | InChI=1S/2C13H21NO3.H2O4S/c2*1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;1-5(2,3)4/h2*4-6,12,14-17H,7-8H2,1-3H3;(H2,1,2,3,4) |
| SMILES | CC(C)(C)NCC(O)c1ccc(O)c(CO)c1.CC(C)(C)NCC(O)c2ccc(O)c(CO)c2.OS(=O)(=O)O |
| IUPAC Name | 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid |
| Molecular Weight (g/mol) | 576.7 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction