missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Albumin (Transport Protein) Mouse, Clone: ALB/2144, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 213-MSM4-P1
This item is not returnable.
View return policy
Specifications
| Albumin (Transport Protein) | |
| Cell Surface and Cytoplasmic | |
| ALB/2144 | |
| Unconjugated | |
| Mouse | |
| 66kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 213 | |
| IgG1 κ |
| ELISA, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human ALB protein | |
| Human serum. Liver or Hepatocellular Carcinoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction