missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Albumin Recombinant Rabbit Monoclonal Antibody (23GB1680), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Albumin |
|---|---|
| Clone | 23GB1680 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088837
![]() |
Invitrogen™
MA554102 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089211
|
Invitrogen™
MA554103 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Albumin Recombinant Monoclonal antibody specifically detects Albumin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Albumin | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P02768, P02770, P07724 | |
| 11657, 213, 24186 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1680 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ALB; ALB protein; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; Bos d 6; bovine serum albumin; BSA; cell growth inhibiting protein 42; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 | |
| A synthesized peptide derived from Cow Bovine Serum Albumin (160-200AA). | |
| Primary | |
| -20°C | |
| ALB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title