missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Albumin and IgG Depletion SpinTrap

Product Code. 10734027 Shop All Cytiva Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
10734027 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10734027 Supplier Cytiva Supplier No. 28948020

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Albumin and IgG Depletion SpinTrap are prepacked columns for the depletion of albumin and IgG from human serum and plasma. They are prepacked with high performance SepharoseTM based media with an affinity for human serum albumin (HSA) and IgG

  • Albumin and IgG Depletion SpinTrap is a prepacked column for the depletion of albumin and IgG from human serum or plasma.
  • Albumin and IgG Depletion SpinTrap handles smaller sample volumes (∼50 μl) using a tabletop centrifuge.
  • Columns are prepacked with high performance Sepharose based media with an affinity for human serum albumin (HSA) and IgG.
  • High depletion capacity, removes > 95% HSA and > 90% IgG, with high reproducibility.
  • Simple and fast procedure.
  • Removal of albumin and IgG allows a higher load of less abundant proteins to be included in the analysis and detection of more proteins.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7 mm
Flow Rate <4 mL/min
Ligand Type Recombinant Protein G fragment and recombinant Protein binding HAS
Particle Size 34 μm
pH 3 to 9
Volume (Metric) 370 μL
Endcapped No
Product Type Albumin and IgG Depletion
compliance-icons
Product Identifier
  • Albumin & IgG Depletion SpinTrap
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.