missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Albumin ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
Description
Albumin is a soluble, monomeric protein with a molecular weight of approximately 65-67 kDa that comprises about one-half of the blood serum protein. Human serum albumin (HAS) is the most abundant protein in human blood plasma, and has a serum half-life of approximately 20 days. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular, unglycosylated serum protein synthesized in the liver as pre-pro-albumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough produce the secreted albumin. Serum albumin has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. The main function of albumin is the regulation of the colloidal osmotic pressure of blood.
Specifications
Specifications
| Accession Number | P02768 |
| Assay Range | 4.915 to 1200 ng/mL |
| Assay Sensitivity | 6 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 213 |
| Gene Symbol | ALB |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction