missing translation for 'onlineSavingsMsg'
Learn More

Albumin, Egg, Powder, Purified, J.T. Baker™

Product Code. p-5391051 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15577834 500 g Poly Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15577834 Supplier Avantor J.T.Baker Supplier No. 044001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 9006-50-2
Molecular Formula C6H8N2O4
Molecular Weight (g/mol) 172.14
InChI Key QCVGEOXPDFCNHA-UHFFFAOYSA-N
Synonym tridodeoylamine
PubChem CID 25022239
IUPAC Name 5,5-dimethyl-2,4-dioxo-1,3-oxazolidine-3-carboxamide
SMILES CC1(C(=O)N(C(=O)O1)C(=O)N)C

Specifications

Packaging Poly Bottle
Quantity 500 g
Grade Purified
Physical Form Powder
Chemical Name or Material Albumin, Egg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.