missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Chemical™ Albumin, Egg, Extra Pure, SLR
Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
Unit Size:
250g
Specifications
Specifications
| Molecular Weight (g/mol) | 33,800 to 46,000g/mol |
| pH Range | 7.6 |
| Quantity | 250 g |
| Packaging | Plastic Powder Jar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction