Learn More
Alfa Aesar™ Albumin, Bovine, New Zealand origin, Low Fatty Acid, Chromatographically Purified
For research use
Brand: Alfa Aesar™ J65400.18
Code : NEW
Additional Details : CAS Number : 9048-46-8 Weight : 0.05000kg
Specifications
Suitable for immunoassays. Reduced fatty acid content minimizes non-specific binding of antibodies | |
Lyophilized powder | |
Bovine Serum | |
RUO | |
No |
Lyophilized Powder | |
50g | |
2° to 8°C | |
Albumin, Bovine, New Zealand origin, Low Fatty Acid, Chromatographically purified |
For Research Use Only