missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Albumin, Bovine, Fraction V Powder
Albumin is used to solubilize lipids, and is also used as a blocking agent in Western blots or ELISA applications.
£2175.00 - £6155.00
Specifications
Source | Bovine Blood |
---|---|
Format | Powder |
Preparation Method | Prior to lyophilization, the product is filtered through a 0.2μm filter |
For Use With (Application) | Immunoassays, Mammalian Cell Culture |
Storage Requirements | 2° to 8°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11475116
|
MP Biomedicals™
08810026 |
5kg |
£6155.00
Each |
Estimated Shipment: 20-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
11465116
|
MP Biomedicals™
08810025 |
1kg |
£2175.00
Each |
Estimated Shipment: 20-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Description
Cream, crystalline “flaky” powder. Prior to lyophilization, the product is filtered through a 0.2mm filter. Low ionic strength and bioburden.
- Solubility: 0.042
- pH: 7.3 (7% solution in 0.15M NaCl)
- Purity: 100%
- Loss on Drying: 3.5%
- Heavy Metals: <10ppm
- Additional Tests: Vesicular Stomatitis Virus: Negative; Bluetongue Virus: Negative
- General: This product was derived from bovine blood collected at a USDA licensed facility. The cattle received ante- and post-mortem health inspection under a veterinarian's supervision and were apparently free from infectious and contagious diseases. All donor animals were sourced from the United States. Veterinary Certificates are maintained on file at the manufacturing site.
Specifications
Bovine Blood | |
Prior to lyophilization, the product is filtered through a 0.2μm filter | |
2° to 8°C |
Powder | |
Immunoassays, Mammalian Cell Culture |