missing translation for 'onlineSavingsMsg'
Learn More

Albumin Antibody (HSA20), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18353641 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353641 0.2 mL 0.20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353641 Supplier Novus Biologicals Supplier No. NBP179049AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Albumin Monoclonal specifically detects Albumin in Human, Bovine (Negative) samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Albumin
Applications Western Blot, ELISA
Classification Monoclonal
Clone HSA20
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbols ALB
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human serum albumin.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 213
Test Specificity No cross-reactivity with other tested human proteins. Does not react with bovine serum albumin, egg white albumin or human alpha fetoprotein.
Target Species Human, Bovine (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.