missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AlbuMax II Lipid Rich BSA

Product Code. 31781125
Change view
Click to view available options
Quantity:
5 g
1 kg
Unit Size:
1kg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31781125 5 g 5g
10624053 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31781125 Supplier Gibco™ Supplier No. 11021011
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤1.0%. Lyophilized powder.

This is a lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤ 1%.

Origin: New Zealand

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 g
Cell Type Mammalian Cells
Product Type BSA
Form Solid

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.