missing translation for 'onlineSavingsMsg'
Learn More

Albendazole, 98+%

Product Code. 11435680
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435680 100 g 100g
11415680 10 g 10g
11425680 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435680 Supplier Thermo Scientific Alfa Aesar Supplier No. H25925.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 54965-21-8 | C12H15N3O2S | 265.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54965-21-8
Molecular Formula C12H15N3O2S
Molecular Weight (g/mol) 265.33
MDL Number MFCD00083232
InChI Key HXHWSAZORRCQMX-UHFFFAOYSA-N
Synonym albendazole, albenza, eskazole, valbazen, zentel, albendazol, proftril, albendazolum, bilutac, zental
PubChem CID 2082
ChEBI CHEBI:16664
IUPAC Name methyl N-(6-propylsulfanyl-1H-benzimidazol-2-yl)carbamate
SMILES CCCSC1=CC=C2N=C(NC(=O)OC)NC2=C1

Specifications

Melting Point 208°C to 210°C
Quantity 100 g
Merck Index 14,210
Formula Weight 265.34
Percent Purity ≥98%
Chemical Name or Material Albendazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.