missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALB Goat anti-Mouse, Polyclonal , Abnova™
Goat polyclonal antibody raised against mouse Alb.
Brand: Abnova PAB29683.1mg
This item is not returnable.
View return policy
Specifications
| ALB | |
| Polyclonal | |
| Goat polyclonal antibody raised against mouse Alb. | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| Alb | |
| Native purified mouse Alb. | |
| 1 mg | |
| Primary | |
| Mouse | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (2 ug/mL) Immunoprecipitation (2 ug/mg) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Alb-1/Alb1 | |
| Goat | |
| Antigen Affinity Purification | |
| RUO | |
| 11657 | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction