missing translation for 'onlineSavingsMsg'
Learn More

ALB-127158(a), MedChemExpress

Product Code. 16475326
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475326 100 mg 100mg
16467167 5 mg 5mg
16460757 50 mg 50mg
16453086 25 mg 25mg
16424448 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16475326 Supplier Medchem Express Supplier No. HY111398
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ALB-127158(a) is a potent and selective melanin concentrating hormone 1 (MCH1) receptor antagonist.

ALB-127158(a) is a potent and selective melanin concentrating hormone 1 (MCH1) receptor antagonist.

Specifications

Chemical Name or Material ALB-127158(a)
CAS 1173154-32-9
Color White
Purity Grade Notes Research
Molecular Formula C23H21FN4O2
Quantity 100 mg
Solubility Information Ethanol : 2 mg/mL (4.95 mM; Need ultrasonic)
SMILES O=C1C=C(OCC2=NC=C(F)C=C2)C=CN1C3=CC4=C(C=C3)C(CNCC5)=C5N4C
Molecular Weight (g/mol) 404.44
Formula Weight 404.44
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.