Learn More
Emma Safety Footwear Alaska Safety Shoes
Product Code. 15858777
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Product Code. 15858777
Brand: Emma Safety Footwear MM30254644
Only null left
Add to basket
Add to basket
Offers exceptional comfort while the steel toe cap protects from falling objects. Easy twist sole provides a 'sticky grip'.
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Certifications/Compliance EN ISO 20345:2011
Color Black
Design Alaska
Liner Material Hydro-Tec™ Sanitized Silver
Material Oiled Nubuck
Size 44