missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALAS2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99389AF350
This item is not returnable.
View return policy
Description
ALAS2 Polyclonal antibody specifically detects ALAS2 in Human samples. It is validated for Western Blot
Specifications
| ALAS2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 212 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 350 | |
| 5-aminolevulinic acid synthase 2,5-aminolevulinate synthase, erythroid-specific, mitochondrial, ALASE, ALAS-E, aminolevulinate, delta-, synthase 2, aminolevulinate, delta-, synthase 2 (sideroblastic/hypochromic anemia), ANH1, ASBXLDPP, Delta-ALA synthase 2, delta-ALA synthetase, Delta-aminolevulinate synthase 2, EC 2.3.1.37, FLJ93603, XLSA | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ALAS2. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction