missing translation for 'onlineSavingsMsg'
Learn More

ALAS2 Antibody (4D8), Novus Biologicals™

Product Code. 18353458 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353458 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353458 Supplier Novus Biologicals Supplier No. H00000212M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ALAS2 Monoclonal antibody specifically detects ALAS2 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALAS2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4D8
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000023
Gene Alias 5-aminolevulinic acid synthase 2,5-aminolevulinate synthase, erythroid-specific, mitochondrial, ALASE, ALAS-E, aminolevulinate, delta-, synthase 2, aminolevulinate, delta-, synthase 2 (sideroblastic/hypochromic anemia), ANH1, ASBXLDPP, Delta-ALA synthase 2, delta-ALA synthetase, Delta-aminolevulinate synthase 2, EC 2.3.1.37, FLJ93603, XLSA
Host Species Mouse
Immunogen ALAS2 (NP_000023, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MVTAAMLLQCCPVLARGPTSLLGKVVKTHQFLFGIGRCPILATQGPNCSQIHLKATKAGGDSPSWAKGHCPFMLSELQDGKSKIVQKAAPEVQEDVKAFK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.