missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALAS1 Recombinant Rabbit Monoclonal Antibody (2A9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538338
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
ALAS-1 is a nuclear-encoded mitochondrial matrix enzyme which catalyzes the condensation of glycine with succinyl-CoA to form delta-amino levulinate, CO2 and CoA. It regulates the first and rate-limiting step of heme biosynthetic pathway. It is one of the two isoforms of ALAS and is a pyridoxal-5' phosphate dependant housekeeping enzyme. It is ubiquitously expressed and is responsible of providing heme for cytochromes and other hemoproteins. ALAS1 in liver undergoes negative feedback regulation by heme. Transcription of ALAS1 gene is stimulated by cAMP and respiratory uncoupling whereas Phorbol esters and insulin repress ALAS1 gene expression. ALAS1 is suggested to be involved in reciprocal regulation of Heme biosynthesis and circadian clock suggesting a potential target for treatment of circadian disorders. ALAS1 gene expression is down regulated in Acute Liver failure resulting in altered heme metabolism and liver function.
Specifications
| ALAS1 | |
| Recombinant Monoclonal | |
| 0.58 mg/mL | |
| PBS with 50% glycerol, 150mM NaCl and 0.02% sodium azide; pH 7.4 | |
| P13196 | |
| ALAS1 | |
| A synthesized peptide derived from human Alas1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 2A9 | |
| Unconjugated | |
| ALAS1 | |
| 5'-aminolevulinate synthase 1; 5-aminolevulinate synthase 1; 5-aminolevulinate synthase, nonspecific, mitochondrial; 5-aminolevulinic acid synthase 1; ALAS; ALA-S; ALAS1; Alas-1; ALAS3; ALASH; Alas-h; ALAS-N; aminolevulinate synthase H; aminolevulinate, delta-, synthase 1; aminolevulinic acid synthase 1; delta-ALA synthase 1; delta-ALA synthetase; delta-aminolevulinate synthase 1; MIG4; migration-inducing protein 4; OK/SW-cl.121; succinyl-CoA: glycine C-succinyl transferase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 211 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction