missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alanyl-tRNA Synthetase Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583712
This item is not returnable.
View return policy
Description
Immunogen sequence: EFTVKNAQVR GGYVLHIGTI YGDLKVGDQV WLFIDEPRRR PIMSNHTATH ILNFALRSVL GEADQKGSLV APDRLRFDFT AKGAMSTQQI KKAEEIANEM IEAAKAVYTQ DCP Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 90%.
The human alanyl-tRNA synthetase (AARS) belongs to a family of tRNA synthases, of the class II enzymes. Class II tRNA synthases evolved early in evolution and are highly conserved. This is reflected by the fact that 498 of the 968-residue polypeptide human AARS shares 41% identity witht the E. coli protein. tRNA synthases are the enzymes that interpret the RNA code and attach specific aminoacids to the tRNAs that contain the cognate trinucleotide anticodons. They consist of a catalytic domain which interacts with the amino acid acceptor-T psi C helix of the tRNA, and a second domain which interacts with the rest of the tRNA structure.
Specifications
| alanyl-tRNA Synthetase | |
| Polyclonal | |
| Unconjugated | |
| AARS1 | |
| Aars; AI316495; alanine tRNA ligase 1, cytoplasmic; alanine--tRNA ligase, cytoplasmic; alanine--tRNA ligase, cytoplasmic {ECO:0000255; Alanyl-tRNA synthetase; alanyl-tRNA synthetase {ECO:0000255; alanyl-tRNA synthetase, cytoplasmic; alaRS; alaRS {ECO:0000255; C76919; CMT2N; EIEE29; HAMAP-Rule:MF_03133}; Protein sticky; Renal carcinoma antigen NY-REN-42; sti | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49588 | |
| AARS1 | |
| Recombinant protein corresponding to Human AARS. Recombinant protein control fragment (Product #RP-104790). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction