missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine-tert-butylester hydrochloride, 98%

Product Code. 10149959 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10149959 5 g Glass Bottle 5g
10351750 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10149959 Supplier Thermo Scientific Acros Supplier No. 434650050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58620-93-2 | C7H16ClNO2 | 181.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58620-93-2
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00058258
InChI Key DOMTZTVJNZKUNX-UHFFFAOYSA-N
Synonym tert-butyl 3-aminopropanoate hydrochloride, h-beta-ala-otbu.hcl, beta-alanine tert-butyl ester hydrochloride, h-beta-ala-otbu hcl, beta-alanine t-butyl ester hydrochloride, 3-amino-propionic acid tert-butyl ester hydrochloride, h-?-ala-otbu.hcl, h-, a-ala-otbu.hcl, h-beta-ala-otbu hydrochloride, tert-butyl 3-aminopropanoate-hcl
PubChem CID 16218890
IUPAC Name tert-butyl 3-aminopropanoate;hydrochloride
SMILES Cl.CC(C)(C)OC(=O)CCN

Specifications

Melting Point 167.8°C to 168.5°C
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 181.66
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material β-Alanine-tert-butylester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.