missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine methyl ester hydrochloride, 98%

Product Code. 10663193 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10663193 5 g Glass Bottle 5g
10591892 25 g Glass Bottle 25g
10175683 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10663193 Supplier Thermo Scientific Acros Supplier No. 371470050
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3196-73-4 | C4H10ClNO2 | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3196-73-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00039060
InChI Key XPGRZDJXVKFLHQ-UHFFFAOYSA-N
Synonym methyl 3-aminopropionate hydrochloride, methyl 3-aminopropanoate hydrochloride, beta-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, h-beta-ala-ome.hcl, h-, a-ala-ome.hcl, h-beta-ala-ome hydrochloride, b-alanine methyl ester hydrochloride, 3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride
PubChem CID 2734767
IUPAC Name methyl 3-aminopropanoate;hydrochloride
SMILES Cl.COC(=O)CCN

Specifications

Melting Point 103.0°C to 105.0°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula NH2CH2CH2COOCH3·HCl
Beilstein 04, IV, 2527
Quantity 5 g
Formula Weight 139.58
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material β-Alanine methyl ester hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.