missing translation for 'onlineSavingsMsg'
Learn More

Alanine (L-Alanine), LoGiCal

Product Code. 30460921
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30460921 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30460921

Brand: LoGiCal LGCFOR0566.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Alanine (L-Alanine)

Specifications

Chemical Name or Material L-Alanine
CAS 56-41-7
Analyte or Component Names L-Alanine
Shipping Condition Room Temperature
Molecular Formula C3 H7 N O2
Certifications/Compliance ISO 9001
Synonym (S)-2-Aminopropanoic Acid, Aspartic Acid Imp. D (Pharmeuropa), L-Alanine, Alanine, L- (7 CI,8 CI), (2 S)-2-Aminopropanoic acid, (S)-(+)-Alanine, (S)-2-Aminopropanoic acid, (S)-Alanine, 2-Aminopropionic acid, Alanine, L-(+)-Alanine, L-2-Aminopropanoic acid, L-2-Aminopropionic acid, Lactamine, NSC 206315, Propanoic acid, 2-amino-, (S)-, L-alpha-Alanine, L-alpha-Aminopropionic acid, alpha-Alanine, alpha-Aminopropionic acid, Aspartic Acid Impurity D, Magnesium Aspartate Dihydrate Impurity E, Magnesium Aspartate Impurity E
InChI Formula InChI=1 S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1
SMILES C[C@H](N)C(=O)O
IUPAC Name (2 S)-2-aminopropanoic acid
Industry Type Forensics
Molecular Weight (g/mol) 89.09
Quantity 10 mg
Formula Weight 89.0477 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.