missing translation for 'onlineSavingsMsg'
Learn More

β-Alanine, ≥98%, Ultrapure, Thermo Scientific™

Product Code. p-7082812 Shop All Thermo Scientific Products
Click to view available options
Quantity:
200 g
1 kg
5 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
200g
5kg
This item is not returnable. View return policy

Product Code. 15895058

Brand: Thermo Scientific J10665.28

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-95-9
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00008200
InChI Key UCMIRNVEIXFBKS-UHFFFAOYSA-N
Synonym beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
PubChem CID 239
ChEBI CHEBI:16958
IUPAC Name 3-aminopropanoic acid
SMILES NCCC(O)=O

Specifications

Melting Point ∽205°C (decomposition)
Packaging Plastic Bottle
Linear Formula NH2CH2CH2COOH
Beilstein 906793
Density 1.437
Quantity 200 g
Formula Weight 89.09
Percent Purity ≥98%
Specific Gravity 1.437
Physical Form Crystalline Powder
Chemical Name or Material beta-Alanine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.