missing translation for 'onlineSavingsMsg'
Learn More

β-Alanine, ≥98%, Ultrapure, Thermo Scientific™

Product Code. 15895058 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 g
1 kg
5 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
200g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15895058 200 g Plastic Bottle 200g
15855248 1 kg Plastic Bottle 1kg
15865248 5 kg Plastic Bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15895058 Supplier Thermo Scientific Supplier No. J10665.28

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-95-9
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00008200
InChI Key UCMIRNVEIXFBKS-UHFFFAOYSA-N
Synonym beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-amino
PubChem CID 239
ChEBI CHEBI:16958
IUPAC Name 3-aminopropanoic acid
SMILES NCCC(O)=O

Specifications

Melting Point ∽205°C (decomposition)
Packaging Plastic Bottle
Linear Formula NH2CH2CH2COOH
Beilstein 906793
Density 1.437
Quantity 200 g
Formula Weight 89.09
Percent Purity ≥98%
Specific Gravity 1.437
Physical Form Crystalline Powder
Chemical Name or Material beta-Alanine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.