missing translation for 'onlineSavingsMsg'
Learn More

Alamethicin

Product Code. 15465989
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465989 1 mg 1mg
15455989 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465989 Supplier Thermo Scientific Alfa Aesar Supplier No. J63829.M^
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A channel-forming ionophore that activates membrane enzymes | CAS: 59588-86-2 | C92H150N22O25 | 1964.341 g/mol

Alamethicin is used as a channel-forming ionophore that activates membrane enzymes, membrane channel-forming peptide. It is a 20-amino acid channel-forming peptide antibiotic that can function as a monovalent cation ionophore. Alamethicin has been used to develop methods for the routine assessment of potential new drug candidates to elicit their pharmacokinetic drug interactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alamethicin is used as a channel-forming ionophore that activates membrane enzymes, membrane channel-forming peptide. It is a 20-amino acid channel-forming peptide antibiotic that can function as a monovalent cation ionophore. Alamethicin has been used to develop methods for the routine assessment of potential new drug candidates to elicit their pharmacokinetic drug interactions.

Solubility
Soluble in DMSO and ethanol.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59588-86-2
Molecular Formula C92H150N22O25
Molecular Weight (g/mol) 1964.341
MDL Number MFCD00151517
InChI Key LGHSQOCGTJHDIL-SLKIUSOBSA-N
Synonym U-22324
PubChem CID 53229968
SMILES CC(C)CC(C(=O)NC(C)(C)C(=O)N1CCCC1C(=O)NC(C(C)C)C(=O)NC(C)(C)C(=O)NC(C)(C)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CC2=CC=CC=C2)CO)NC(=O)CNC(=O)C(C)(C)NC(=O)C(C(C)C)NC(=O)C(C)(C)NC(=O)C(CCC(=O)N)NC(=O)C(C)NC(=O)C(C)(C)NC(=O)C(C)NC(=O)C(C)(C)NC(=O)C3CCCN3C

Specifications

Type Alamethicin
Physical Form Powder
Melting Point 267°C to 270°C
Color White
Quantity 1 mg
UN Number UN3462
Solubility Information Soluble in DMSO and ethanol.
Formula Weight 1964.4
Chemical Name or Material Alamethicin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.