missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ alamarBlue™ Dye

Product Code. p-4525225
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 15549604

Brand: Thermo Scientific™ Y00025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allows three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.

Thermo Scientific™ alamarBlue provides unmatched sensitivity and versatility, allowing three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.

alamarBlue, a resazurin-based product with extended incubation time, is stable for up to 5 days.
TRUSTED_SUSTAINABILITY

Specifications

Description alamarBlue indicator dye
Product Type Indicator Dye
Quantity 25 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.