missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ alamarBlue™ Dye

Product Code. 15549604 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15549604 25 mL 25mL
15559604 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15549604 Supplier Thermo Scientific™ Supplier No. Y00025
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Allows three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.

Thermo Scientific™ alamarBlue provides unmatched sensitivity and versatility, allowing three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.

alamarBlue, a resazurin-based product with extended incubation time, is stable for up to 5 days.
TRUSTED_SUSTAINABILITY

Specifications

Description alamarBlue indicator dye
Product Type Indicator Dye
Quantity 25 mL
How do alamarBlue reagent and PrestoBlue reagent differ from resazurin and C12-resazurin?

alamarBlue reagent and PrestoBlue reagent contain resazurin in a proprietary stabilizing formulation that allows for a convenient “mix, incubate, and read” protocol. PrestoBlue reagent is an improvement in the formulation of alamarBlue reagent that allows for much faster staining (typically 10 minutes vs. 1-4 hours to obtain a similar signal and sensitivity). C12-resazurin is a derivative of resazurin that has better cellular retention and thus allows for analysis on a flow cytometer and multiplexing with viability indicators and other biomarkers.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.