Learn More
Thermo Scientific™ alamarBlue™ Cell Viability Assay Reagent
Product Code. 13474309 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13474309 25 mL 25mL
13494309 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13474309 Supplier Thermo Scientific™ Supplier No. 88951
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Utilize a redox-sensitive growth indicator to quantitatively measure cell viability and proliferation using fluorescent or colorimetric detection strategies.
Utilize a redox-sensitive growth indicator to quantitatively measure cell viability and proliferation using fluorescent or colorimetric detection strategies with Thermo Scientific™ alamarBlue Cell Viability Assay Reagent.
Additional Information and Online Ordering
The alamarBlue Reagent is a non–toxic, water-soluble resazurin dye that yields a fluorescent signal and a colorimetric change when incubated with metabolically active cells. The reagent is designed for cell viability and proliferation studies using mammalian cell lines, bacteria and fungi and is ideally suited for in vitro cytotoxicity studies. Additionally, cell viability can be monitored in real time when performing a variety of tissue engineering applications.
Highlights:
  • High sensitivity and linearity – reproducibly detect as few as 50 cells
  • Quantitative – provides accurate measurement over time
  • Non-toxic to cells – less likely to interfere with normal metabolism
  • Scalable – mix–and–read, homogeneous assay enhances speed while minimizing effort
  • No cell lysis – ideal for use with time course experiments or post–measurement functional assays
  • Stable – allows for continuous cell growth monitoring over a long period
  • Flexible – can be used with primary cell lines, adherent or suspension cells, bacteria and fungi
  • Versatile – allows fluorescent or colorimetric detection methods
Requires:
Fluorescent plate reader for excitation at 530 to 560nm and emission detection at 590nm, or spectrophotometer plate reader capable of reading 570nm and 600nm absorbance.
Recommended for:
  • Monitor cell proliferation and cell viability for in vitro cytotoxicity studies (e.g., effects of environmental toxins, cytokines, drugs, siRNA/microRNA)
  • Monitor cell viability in real time for 3D tissue engineering applications
TRUSTED_SUSTAINABILITY
Specifications
Description alamarBlue Cell Viability Assay Reagent
Quantity 25 mL
Sufficient For 2500 reactions in a 96-well plate or 10,000 reactions in a 384-well plate