missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALADIN Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15127-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the WD-repeat family of regulatory proteins and may be involved in normal development of the peripheral and central nervous system. The encoded protein is part of the nuclear pore complex and is anchored there by NDC1. Defects in this gene are a cause of achalasia-addisonianism-alacrima syndrome (AAAS), also called triple-A syndrome or Allgrove syndrome. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| ALADIN | |
| Polyclonal | |
| Unconjugated | |
| AAAS | |
| AAA, AAAS, AAASb, ADRACALA, ADRACALIN, ALADIN, GL003 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8086 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NRG9 | |
| AAAS | |
| ALADIN Fusion Protein Ag7211 | |
| 150 μL | |
| Primary | |
| Human, Non-human Primate | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction