missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Zn/Mg/Cu, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Aluminum, Al/Zn/Mg/Cu, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Cadmium/Nickel/Vanadium/Chromium/Lead/Zirconium/Titanium/Tin/Manganese/Silicon/Copper/Iron/Magnesium/Zinc |
| CAS | 7429-90-5 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cadmium 0.00054 %/Nickel 0.0037 %/Vanadium 0.0067 %/Chromium 0.071 %/Lead 0.073 %/Zirconium 0.08 %/Titanium 0.119 %/Tin 0.219 %/Manganese 0.224 %/Silicon 0.59 %/Copper 0.81 %/Iron 1.04 %/Magnesium 1.47 %/Zinc 5.3 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction