missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Cu, AA 2007, A92007, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Aluminum, Al/Cu, AA 2007, A92007, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Boron/Zirconium/Calcium/Nickel/Titanium/Chromium/Tin/Bismuth/Iron/Silicon/Zinc/Manganese/Magnesium/Lead/Copper |
| CAS | 7429-90-5 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0018 %/Zirconium 0.0021 %/Calcium 0.0051 %/Nickel 0.0087 %/Titanium 0.0236 %/Chromium 0.0352 %/Tin 0.0676 %/Bismuth 0.147 %/Iron 0.252 %/Silicon 0.328 %/Zinc 0.386 %/Manganese 0.605 %/Magnesium 0.663 %/Lead 0.889 %/Copper 3.699 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction